Learn More
Medchemexpress LLC Z-D-Asp-OH | 78663-07-7 | C12H13NO6 | 500 G

Supplier: Medchemexpress LLC HY77757500G
Z-D-Asp-OH is an aspartic acid derivative primarily used as a laboratory chemical and in the manufacture of various substances. As an amino acid derivative, it influences the secretion of anabolic hormones, contributes to the supply of fuel during exercise, and supports mental performance during stress-related tasks. It also helps prevent exercise-induced muscle damage. This product is intended for research use only by experienced personnel.
- Aspartic acid derivative
- Influences secretion of anabolic hormones
- Supplies fuel during exercise
- Enhances mental performance during stress-related tasks
- Prevents exercise-induced muscle damage
- Stable as powder at -20°C for 3 years or 4°C for 2 years
- Stable in solvent at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.