Learn More
STA PHARMACEUTICAL US LLC Z-D-beta-Dab(Fmoc)-OH | 5 g | CAS 349547-09-7 | MDL MFCD03788645
Supplier: STA PHARMACEUTICAL US LLC STAAA104215GM
Z-D-beta-Dab(Fmoc)-OH is a Amino Acid reagent (Subcategory: Beta AA) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 349547-09-7
- MDL: MFCD03788645
- InChIKey: GNBVOVLCIBZKIQ-LJQANCHMSA-N
- Molecular Weight: 474.513
- Molecular Formula: C27H26N2O6
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (R)-4-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(((benzyloxy)carbonyl)amino)butanoic acid
- SMILES: OC(C[C@@H](NC(OCC1=CC=CC=C1)=O)CNC(OCC2C3=CC=CC=C3C4=CC=CC=C24)=O)=O
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.