Learn More
Medchemexpress LLC Z-D-Dab(Boc)-OH dicyclohexylamine | 214852-61-6 | 97.0% | 533.7 g/mol | C29H47N3O6 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW090421
Encompass_Preferred
Z-D-Dab(Boc)-OH dicyclohexylamine is a protected amino-acid derivative (the dicyclohexylamine salt of Z-D-2,4-diaminobutyric acid) used as a biochemical assay reagent and peptide synthesis building block. It is supplied in small laboratory pack sizes for research use only.
  • Protected D-2,4-diaminobutyric acid derivative suitable for peptide synthesis.
  • Provided as the dicyclohexylamine salt for improved handling.
  • Available in multiple pack sizes, including 250 mg.
  • Molecular formula C29H47N3O6 and CAS number 214852-61-6.
  • Typical distributor-reported purity around 97.0% (varies by supplier).
Catalog No. 50-002-09528