Learn More
Medchemexpress LLC Z-DEVD-AFC | 1135416-11-3 | 98.1% | 821.71 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP19861MG
Encompass_Preferred
Z-DEVD-AFC is a cell-permeant substrate for caspase-3. It induces a shift in fluorescence upon cleavage of the AFC fluorophore and can be utilized to detect caspase-3-like enzyme activity.
  • Cell-permeant substrate for caspase-3
  • Causes a shift in fluorescence upon cleavage of the AFC fluorophore
  • Detects caspase-3-like enzyme activity
  • Fluorescent dye with excitation at 380 nm and emission at 500 nm
  • Protease substrate
Catalog No. 50-004-23971