Learn More
Selleck Chemical LLC Z-DEVD-FMK 1mg 210344-95-9 Caspase-3 Inhibitor
SDP
Supplier:  Selleck Chemical LLC S73121MG
Encompass_Preferred
Z-DEVD-FMK (Caspase-3 Inhibitor) is a specific, irreversible Caspase-3 inhibitor, and also shows potent inhibition on caspase-6, caspase-7, caspase-8, and caspase-10. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1057386