Learn More
Apexbio Technology LLC Z-DEVD-pNA 5mg
SDP
Supplier:  Apexbio Technology LLC A87485
Encompass_Preferred
Z-DEVD-pNA is a colorimetric peptide substrate targeting caspase-3 It is designed to enable detection and quantification of caspase-3 enzymatic activity thereby facilitating the assessment of apoptosis Z-DEVD-pNA exerts its biological function through cleavage by active caspase-3 resulting in the release of p-nitroaniline which can be measured spectrophotometrically at 400 nm Based on these properties Z-DEVD-pNA holds research potential in apoptosis detection and analysis studies
Catalog No. 50-000-93876