Learn More
Selleck Chemical LLC Z-FA-FMK
SDP
Supplier:  Selleck Chemical LLC S73915MG
Encompass_Preferred
Z-FA-FMK ((1S)-Z-FA-FMK) is an irreversible cysteine protease inhibitor and also inhibits effector caspases
Catalog No. 50-000-01453