Learn More
Medchemexpress LLC (Z)-FeCP-oxindole | 1137967-28-2 | 99.6% | 329.17 | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HY10844410MG
Encompass_Preferred
(Z)-FeCP-oxindole is a selective human vascular endothelial growth factor receptor 2 (VEGFR2) inhibitor with an IC50 value of 200 nM. It can significantly inhibit VEGFR1 and PDGFRa or b at 10 μM. (Z)-FeCP-oxindole also exhibits anticancer activity, acting on B16 murine melanoma lines with an IC50 less than 1 μM.
  • Selective human vascular endothelial growth factor receptor 2 (VEGFR2) inhibitor
  • Inhibits VEGFR1 and PDGFRa or b
  • Exhibits anticancer activity
Catalog No. 50-003-45457