Learn More
FUJIFILM BIOSCIENCES INC Z-Ferimzone Standard
Supplier:  FUJIFILM BIOSCIENCES INC 06604121
Encompass_Preferred
Z-Ferimzone Standard is a highly pure crystalline analytical reference material used in pesticide residue analysis Validated using both HPLC and quantitative NMR to ensure quality it is ideal for use in regulatory and quality assurance environments Requires cool dark storage in a sealed container
Catalog No. 50-004-50389