Learn More
Medchemexpress LLC (Z)-flunarizine | 693765-11-6 | 100mg
SDP
Supplier:  Medchemexpress LLC HYZ8176100MG
Encompass_Preferred
(Z)-Flunarizine is the (Z)-isomer of Flunarizine (HY-B0358) Flunarizine is a potent dual Na /Ca2 channel (T-type) blocker Flunarizine is a D2 dopamine receptor antagonist Flunarizine shows anticonvulsive and antimigraine activity and peripheral vasodilator effects[1]
Catalog No. 50-002-68914