Learn More
Medchemexpress LLC Z-Glu(OBzl)-OH | 5680-86-4 | 99.06% | 371.38 | 25 G

Supplier: Medchemexpress LLC 25GHYW011074
Z-Glu(OBzl)-OH is a glutamic acid derivative. Amino acids and amino acid derivatives are commercially used as ergogenic supplements. They influence the secretion of anabolic hormones, supply fuel during exercise, improve mental performance during stress-related tasks, and prevent exercise-induced muscle damage. These compounds are recognized as beneficial ergogenic dietary substances.
- Molecular weight: 371.38
- Formula: C20H21NO6
- CAS no.: 5680-86-4
- Appearance: Solid
- Color: White to off-white
- Shipping: Room temperature in continental US
- Storage: Powder stored at -20°C for 3 years, 4°C for 2 years; in solvent at -80°C for 6 months, -20°C for 1 month
- Solubility (in vitro): Soluble in DMSO at 100 mg/mL (269.27 mM) with sonication
- Purity: 99.06%
- Application: For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.