Learn More
Medchemexpress LLC Z-Gly-Arg-Thiobenzyl ester | 130698-88-3 | 98.6% | 471.57 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP527810MG
Encompass_Preferred
Z-Gly-Arg-Thiobenzyl ester is a chromogenic substrate specifically designed for plasmogen-activated serine proteases. This compound is intended strictly for research purposes and is not for direct use with patients.
  • Chromogenic substrate
  • For plasmogen-activated serine proteases
  • Intended for research use only
  • Not for sale to patients
Catalog No. 50-003-98320