Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Z-Gly-Arg-thiobenzyl Ester, MP Biomedicals™

Z-Gly-Arg-thiobenzyl Ester is a chromogenic substrate for Plasmogen Activator Serine Protease.

$434.00 - $695.00

Specifications

Form Semi-solid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
03SB00705
View Documents
MP Biomedicals, Inc
03SB00705
5 mg
Each for $434.00
Add to Cart
 
03SB00710
Accessories View Documents
MP Biomedicals, Inc
03SB00710
10 mg
Each for $695.00
Add to Cart
 
Description

Description

  • Peptide thiobenzyl esters are sensitive substrates for serine and cysteine proteases.
  • The thiol leaving group is detected by reaction with a thiol reagent (DTNB or Leman's reagent) or 4,4'-dithiodipyridine.
  • The rate of thiobenzyl ester hydrolysis is measured at 324 nm by monitoring the formation of 4-thiopyridone (E324nm = 19,800 M-1cm-1) or at 405 nm using DTNB as a thiol reagent (E405nm = 13,260 M-1cm-1) for 2-nitro-5-thiobenzoate anion TNBS.
  • Protein or Enzyme Type: Activators, Inhibitors and Substrates
Specifications

Specifications

Semi-solid
Videos
SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.