Learn More
Bachem Z-Gly-Gly-Gly-OSu
SDP
Supplier:  Bachem 4001727.0001
Encompass_Preferred
Z-Gly-Gly-Gly-OSu, 138346-57-3, C18H20N4O8, Mr 420.38, 1g. NHS active ester of Z-triglycine.
Catalog No. 50-313-087