Learn More
Bachem Z-Gly-Pro-AMC
SDP
Supplier:  Bachem 4002518.0050
Encompass_Preferred
Z-Gly-Pro-AMC, 68542-93-8, C25H25N3O6, Mr 463.49, 50mg. Z-GP-AMC, fluorogenic substrate for the determination of post-proline cleaving enzyme (prolyl endopeptidase).
Catalog No. NC1103032