Learn More
Bachem Z-Gly-Pro-Arg-pNA
SDP
Supplier:  Bachem 4000768.0100
Encompass_Preferred
Z-Gly-Pro-Arg-pNA Acetate, 66648-35-9, C27H34N8O7, Mr 582.62, 100mg. Z-GPR-pNA. Z-GPR-pNA, a chromogenic serine protease substrate. Kinetic data for Thrombin: kcat = 127, Km = 4.18.
Catalog No. 50-260-412