Learn More
Medchemexpress LLC Z-Gly-Pro-pNA | 65022-15-3 | 99.4% | 426.42 | 50 MG

Supplier: Medchemexpress LLC HY13722050MG
Z-Gly-Pro-pNA serves as a substrate for measuring prolyl endopeptidase (PEP) inhibitory activity.
- High purity of 99.4%
- Solid appearance
- White to off-white color
- Molecular weight of 426.42
- Chemical formula: C21H22N4O6
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.