Learn More
Bachem Z-Gly-Pro-Arg-AMC · HCl
SDP
Supplier:  Bachem 4002047.0025
Encompass_Preferred
Z-Gly-Pro-Arg-AMC · HCl, 201928-42-9, C31H37N7O7 · HCl, Mr 656.14, 25mg. Z-GPR-AMC is a fluorogenic substrate for cathepsin K, granzyme A, and thrombin. Kinetic data for throbin: kcat = 18.6, Km = 21.7 uM.
Catalog No. 50-194-7024