Learn More
TARGETMOL CHEMICALS INC Z-GUGGULSTERONE 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1728010MG
Encompass_Preferred
Also available in 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (Z)-Guggulsterone inhibits the growth of human prostate cancer cells by causing apoptosis. Z-guggulsterone inhibits angiogenesis by suppressing the VEGF–VEGF-R2–Akt signaling axis. purity: 99%
Catalog No. 50-257-7312