Learn More
Enzo Life Sciences Z-IE(OMe)TD(OMe)-FMK (1mg)
SDP
Supplier:  Enzo Life Sciences ALX260073M001
Encompass_Preferred
Irreversible, cell permeable inhibitor of caspase-8 and granzyme B. Inhibits also caspase-10. Alternative name: Caspase 8 inhibitor (fluoromethylketone), Granzyme B inhibitor (fluoromethylketone). Formula: C30H43FN4O11. MW: 654.7. Purity: ≥95% (TLC). Appearance: White to off-white solid. Solubility: Soluble in DMSO. Long Term Storage: -20°C. Use/Stability: Store stock solutions in aliquots at -20°C. Handling: Packaged under inert gas.
Catalog No. 50-200-8286