Learn More
Medchemexpress LLC Z-IE(OMe)TD(OMe)-FMK | 210344-98-2 | 98.0% | 654.68 g/mol | C30H43FN4O11 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10129725MG
Encompass_Preferred
Z-IETD-FMK is a synthetic peptide-based, cell-permeable inhibitor of caspase-8 that also inhibits granzyme B. It is supplied for in vitro research use to study apoptosis and protease-mediated signaling.
  • Selective caspase-8 and granzyme B inhibition.
  • Cell-permeable peptide-based inhibitor suitable for cellular assays.
  • High purity: 98.0%.
  • Molecular formula: C30H43FN4O11; molecular weight: 654.68 g/mol.
  • Supplied in milligram-scale quantities for laboratory research.
Catalog No. 50-002-12777