Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Z-IETD-AFC | 219138-02-0 | 99.7% | 821.75 | 1 MG
SDP

Supplier:  Medchemexpress LLC HYP22081MG

Encompass_Preferred

Z-IETD-AFC is a specific fluorescence substrate utilized to determine caspase-8 catalytic activity. This product is intended for research use only.

  • Specific fluorescence substrate for caspase-8.
  • Can be used to determine caspase-8 catalytic activity.
  • High purity (99.74%).
  • Store sealed, away from moisture and light, under nitrogen.
  • Powder storage: -80°C for 2 years or -20°C for 1 year.
  • Storage in solvent: -80°C for 6 months or -20°C for 1 month (sealed, away from moisture and light, under nitrogen).

Catalog No. 50-004-22277


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.