Learn More
Medchemexpress LLC Z-IETD-AFC | 219138-02-0 | 99.7% | 821.75 | 5 MG

Supplier: Medchemexpress LLC HYP22085MG
Z-IETD-AFC is a specific fluorescence substrate designed to determine caspase-8 catalytic activity.
- Specific fluorescence substrate
- Determines caspase-8 catalytic activity
- Purity of 99.74%
- Molecular weight of 821.75
- Chemical formula: C37H42F3N5O13
- Appears as a white to off-white solid
- Emission wavelength: 500
- Excitation wavelength: 380
- Soluble in DMSO: ≥ 100 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.