Learn More
Selleck Chemical LLC Z-IETD-FMK
SDP
Supplier:  Selleck Chemical LLC S731410MM/1ML
Encompass_Preferred
Z-IETD-FMK (Caspase-8 Inhibitor Z-IE(OMe)TD(OMe)-FMK) is a specific Caspase-8 inhibitor Z-IETD-FMK is also an inhibitor of granzyme B
Catalog No. 50-000-04356