Learn More
Medchemexpress LLC Z-IETD-FMK (Z-IE(OMe)TD(OMe)-FMK) | 210344-98-2 | MFCD03490491 | >98.0% | 654.68 g/mol | C30H43FN4O11 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10129750MG
Encompass_Preferred
Z-IETD-FMK (Z-IE(OMe)TD(OMe)-FMK) is a cell-permeable, selective caspase-8 inhibitor commonly used in apoptosis and caspase pathway research. It also exhibits activity against granzyme B and is supplied at research-scale purities suitable for in vitro studies.
  • Selective and cell-permeable caspase-8 inhibitor.
  • Also inhibits granzyme B, enabling broader apoptosis pathway studies.
  • High purity (>98.0%) suitable for in vitro research applications.
  • Available in multiple research-scale package sizes, including 50 MG.
  • Peptide sequence: Z-Ile-Glu-Thr-Asp-FMK.
Catalog No. 50-002-11288