Learn More
Medchemexpress LLC Z-Ile-Glu(OMe)-Thr-Asp(OMe)-fluoromethylketone | 210344-98-2 | ≥98.0% | 654.68 | C30H43FN4O11 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10129710MG
Encompass_Preferred
Z-IETD-FMK is an irreversible, cell-permeable peptide fluoromethylketone inhibitor selective for caspase-8 that also inhibits granzyme B. It is used to block caspase-8-dependent apoptotic signaling in biochemical and cell-based assays and is supplied as a lyophilized powder with established solubility and storage recommendations.
Catalog No. 50-002-12776