Learn More
Bachem Z-Ile-Glu(OMe)-Thr-DL-Asp(OMe)-fluoromethylketone
SDP
Supplier:  Bachem 4034771.0001
Encompass_Preferred
Z-Ile-Glu(OMe)-Thr-DL-Asp(OMe)-fluoromethylketone, 1926163-59-8, C30H43FN4O11, Mr 654.69, 1mg. Z-IETD-FMK. Cell permeable inhibitor of caspase-8 and granzyme B. The bis-methyl ester of Z-IETD-FMK blocked CD95-induced apoptosis.
Catalog No. 50-260-567