Learn More
Medchemexpress LLC (Z)-Indirubin | 906748-38-7 | 262.26 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0117A100MG
Encompass_Preferred
(Z)-Indirubin is an isomer of Indirubin, a bis-indole alkaloid that has demonstrated anticancer activity against chronic myelocytic leukemia. This product is intended for research use only.
  • Isomer of indirubin
  • Bis-indole alkaloid
  • Exhibits anticancer activity against chronic myelocytic leukemia
  • Suitable for research use
Catalog No. 50-004-30282