Learn More
Medchemexpress LLC HY-107842 5mg Medchemexpress, (Z)-JIB-04 CAS:199596-24-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1078425MG
Encompass_Preferred
Medchemexpress, HY-107842 5mg (Z)-JIB-04 CAS:199596-24-2 (Z)-JIB-04 (NSC693627) is the Z isomer of JIB-04 that has two forms, E (HY-13953) and Z isomers. (Z)-JIB-04 is inactive in epigenetic analysis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9611