Learn More
Medchemexpress LLC (Z)-JQ1-TCO | 2087490-43-3 | 99.3% | 609.18 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY148864A
Encompass_Preferred
JQ1-TCO (JQ1-trans-cyclooctene) is a derivative of JQ1 (HY-13030), which is an inhibitor of BET. It is suitable for click chemistry and can be used as a molecular probe both in vitro and in vivo.
  • Suitable for click chemistry.
  • Can be used as molecular probes in vitro and in vivo.
Catalog No. 50-003-63525