Learn More
Medchemexpress LLC (Z)-JQ1-TCO | 2087490-43-3 | 99.3% | 609.18 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY148864A5MG
Encompass_Preferred
JQ1-TCO, a derivative of JQ1, functions as an inhibitor of BET. This compound is suitable for click chemistry applications and can be utilized as molecular probes in various research settings.
  • Suitable for click chemistry
  • Functions as an inhibitor of BET
  • Used as molecular probes in vitro
  • Used as molecular probes in vivo
Catalog No. 50-003-59849