Learn More
Medchemexpress LLC (Z)-JQ1-TCO | 2087490-43-3 | 99.3% | 609.18 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY148864A50MG
Encompass_Preferred
JQ1-TCO (JQ1-trans-cyclooctene) is a derivative of JQ1, an inhibitor of BET. This compound is suitable for click chemistry and can be used as molecular probes in vitro and in vivo.
  • Suitable for click chemistry
  • Can be used as molecular probes in vitro
  • Can be used as molecular probes in vivo
Catalog No. 50-003-61529