Learn More
Cayman Chemical Z- L-Arg-AMChydrochlorIde 25mg
SDP
Supplier:  Cayman Chemical 3079725 MG
Encompass_Preferred
A fluorogenic substrate for trypsin, cathepsin B, and cathepsin H; AMC is released upon enzymatic cleavage by trypsin, cathepsin B, or cathepsin H and its fluorescence can be used to quantify trypsin, cathepsin B, and cathepsin H activity; ex/em = 340-360/440-460 nm, respectively
Catalog No. 50-298-9086