Learn More
Medchemexpress LLC Z-L-Val-OH | 1149-26-4 | MFCD00008922 | 99.79% | 251.28 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW00875350G
Encompass_Preferred
Z-L-Val-OH is an amino acid derivative that serves as a key intermediate in organic synthesis. It is utilized in the synthesis of anti-cytomegalovirus compounds and peptide inhibitors of human spleen plasminogen activator (SFP) and human leukocyte elastase-like protease (ELP).
  • An amino acid derivative
  • Key intermediate in organic synthesis
  • Used in the synthesis of anti-cytomegalovirus compounds
  • Applied in peptide inhibitors of human spleen plasminogen activator (SFP)
  • Utilized in peptide inhibitors of human leukocyte elastase-like protease (ELP)
Catalog No. 50-004-51275