Learn More
TARGETMOL CHEMICALS INC Z-LEHD-fmk 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T218351MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Z-LEHD-FMK is a selective and potent caspase-9 inhibitor that protects against reperfusion injury and slows apoptosis. Z-LEHD-FMK exhibits antitumor and neuroprotective activity, increases the yield of in vitro-produced preimplantation embryos of [Bubalus bubalis], and alters the cellular stress response. Purity 96.13%
Catalog No. 50-287-9483