Learn More
Medchemexpress LLC Z-LEHD-FMK | 210345-04-3 | 98.1% | 690.72 | 5 MG

Supplier: Medchemexpress LLC HYP10105MG
Z-LEHD-FMK is a selective and irreversible inhibitor of caspase-9. It protects against lethal reperfusion injury and attenuates apoptosis, also demonstrating a neuroprotective effect in a rat model of spinal cord trauma.
- Selective and irreversible caspase-9 inhibitor
- Protects against lethal reperfusion injury
- Attenuates apoptosis
- Neuroprotective in spinal cord trauma models
- Purity: 98.1%
- Molecular Weight: 690.72
- Appearance: Solid, white to yellow
- Solubility: 100 mg/mL in DMSO
- Storage (powder): -80°C for 2 years, -20°C for 1 year
- Storage (in solvent): -80°C for 6 months, -20°C for 1 month (sealed, away from moisture)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.