Learn More
Medchemexpress LLC Z-LEHD-FMK | 210345-04-3 | 98.1% | 690.72 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYP101050MG
Encompass_Preferred
Z-LEHD-FMK is a selective and irreversible inhibitor of caspase-9. It protects against lethal reperfusion injury, attenuates apoptosis, and demonstrates neuroprotective effects in a rat model of spinal cord trauma. This compound is a potent tool for research into apoptosis and neuroprotection.
  • Selective and irreversible inhibitor of caspase-9
  • Protects against lethal reperfusion injury
  • Attenuates apoptosis
  • Neuroprotective effect in spinal cord trauma models
Catalog No. 50-004-24625