Learn More
Carbosynth LLC. Z-Leu-Arg-Gly-Gly-MCA (5 mg vial), 1ea, MW: 692.76, Molecular formula: C34H44N8O8, CAS# 167688-68-2
SDP
Supplier:  Carbosynth LLC. MLG3176V5MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: Benzyloxycarbonyl-L-Leucyl-L-Arginyl-Glycyl-Glycine 4-Methyl-Coumaryl-7-Amide, Z-Leu-Arg-Gly-Gly-AMC, Z-LRGG-AMC, Z-LRGG-MCA, Purity: , Grade A, Application: Substrate for Isopeptidase, Storage Temperature: -20C, References: T R.L Stein, Z. Chen, and F. Melandri, Biochemistry, 34, 12616 (1995).
Catalog No. 50-168-5988