Learn More
Carbosynth LLC. Z-Leu-Leu-H (aldehyde) (5 mg vial), 1ea, MW: 362.46, Molecular formula: C20H30N2O4, CAS# 26305-03-3
SDP
Supplier:  Carbosynth LLC. IZL3178V5MG
Encompass_Preferred
This item has a minimum order quantity of 3 per supplier requirements
Synonyms: Benzyloxycarbonyl-L-Leucyl-L-Leucinal; Cbz-Leu-Leu-CHO, Z-Leu-Leu-al, Application: Calpain inhibitor, Storage Temperature: -20C, References: Y. Saito, S. Tsubuki, H. Ito, and S. Kawashima, Neurosci. Lett., 120, 1 (1990)., S. Tsubuki, Y. Saito, M. Tomioka, H. Ito, and S. Kawashima, J. Biochem., 119, 572 (1996).
Catalog No. 50-168-5912