Learn More
Apexbio Technology LLC Z-LEVD-FMK 1135688-25-3 5mg
SDP
Supplier:  Apexbio Technology LLC A90465
Encompass_Preferred
Z-LEVD-FMK (CAS 1135688-25-3) is a cell-permeable inhibitor that selectively targets caspase-4 by covalently binding to its active site thereby preventing enzymatic activity By blocking caspase-4 Z-LEVD-FMK interferes with the endoplasmic reticulum (ER) stress-mediated apoptosis pathway This compound is widely utilized in studies investigating the molecular mechanisms of apoptosis particularly those examining the role of ER stress-induced signaling in tumor cell death It serves as a valuable tool for elucidating caspase-4 function in cellular stress responses and programmed cell death
Catalog No. 50-000-93909