Learn More
Medchemexpress LLC Z-LEVD-FMK | 1135688-25-3 | 98.0% | 100 MG

Supplier: Medchemexpress LLC HY128707100MG
Z-LEVD-FMK is a cell-permeable caspase-4 inhibitor that effectively blocks ER stress-induced apoptosis in cancer cells.
- Cell-permeable caspase-4 inhibitor
- Blocks ER stress-induced apoptosis in cancer cells
- Purity of 98.0%
- Molecular weight: 652.71
- Chemical formula: C31H45FN4O10
- Appearance: solid, white to off-white
- Soluble in DMSO at 125 mg/mL
- Powder storage: -20°C for 3 years, 4°C for 2 years
- In-solvent storage: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.