Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Z-LEVD-FMK | 1135688-25-3 | 98.0% | 50 MG
SDP

Supplier:  Medchemexpress LLC HY12870750MG

Encompass_Preferred

Z-LEVD-FMK is a cell-permeable caspase-4 inhibitor that blocks ER stress-induced apoptosis in cancer cells. It has a molecular weight of 652.71 and a chemical formula of C31H45FN4O10, appearing as a white to off-white solid. It can be dissolved in DMSO at 125 mg/mL.

  • Cell-permeable caspase-4 inhibitor
  • Blocks ER stress-induced apoptosis in cancer cells
  • Molecular weight of 652.71
  • Chemical formula C31H45FN4O10
  • Appears as a white to off-white solid
  • Dissolves in DMSO at 125 mg/mL
  • Blocks IL-1β-induced IL-8 production in hRPE cells
  • Inhibits caspase-3 activity in hRPE cells
  • Completely blocks E2-induced PARP cleavage in 5C cells
  • Reverses E2-inhibited growth and prevents morphologic alterations associated with apoptosis in 5C cells

Catalog No. 50-003-42009


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.