Learn More
Medchemexpress LLC Z-Lys-OMe hydrochlo 500mg
SDP
Supplier:  Medchemexpress LLC HYW098322500MG
Encompass_Preferred
Z-Lys-OMe hydrochloride is a synthetic intermediate that can be used in the synthesis of peptide enzyme substrates and lysine cationic surfactants
Catalog No. 50-002-05273