Learn More
Medchemexpress LLC Z-Lys-OMe hydrochlo 5g
SDP
Supplier:  Medchemexpress LLC HYW0983225G
Encompass_Preferred
Z-Lys-OMe hydrochloride is a synthetic intermediate that can be used in the synthesis of peptide enzyme substrates and lysine cationic surfactants
Catalog No. 50-002-06155