Learn More
Apexbio Technology LLC Z-Lys(Z)-OH 405-39-0 100g
SDP
Supplier:  Apexbio Technology LLC A6953100000
Encompass_Preferred
Z-Lys(Z)-OH (CAS 405-39-0) is a lysine derivative containing benzyloxycarbonyl (Z) protecting groups designed for selective use in peptide and protein synthetic procedures The controlled and selective removal of its protecting groups enables targeted chemical modifications and stepwise elongation in both solid-phase and solution-phase peptide synthesis protocols Z-Lys(Z)-OH is often employed in experimental studies examining substrate specificity mechanisms of proteolytic enzymes protein modification pathways and in designing peptide analogs for probing structure-function relationships Additionally it serves as a research reagent for synthesizing defined peptide structures facilitating investigation of their biological activities and mechanistic roles in biochemical systems
Catalog No. 50-000-94979