Learn More
Medchemexpress LLC Z-Lys(Z)-OSu | 21160-83-8 | MFCD00057964 | >98.0% | 511.52 g·mol⁻1 | C26H29N3O8 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW1295875G
Encompass_Preferred
Z-Lys(Z)-OSu is a carbamate-protected lysine N-hydroxysuccinimide ester used as an activated amino acid derivative for peptide coupling and site-specific lysine modification. The reagent combines benzyloxycarbonyl (Z) protection with an OSu leaving group to enable efficient amide bond formation in peptide synthesis and bioconjugation workflows.
  • Activated N-hydroxysuccinimide (OSu) ester for efficient coupling.
  • Benzyloxycarbonyl (Z) protection on α- and ε-amines to control selectivity.
  • Suitable for peptide synthesis and lysine-specific derivatization.
  • High purity for consistent reaction performance.
  • Stable as a dry powder under low-temperature storage.
  • Soluble in common organic solvents for reaction use.
Catalog No. 50-002-83433