Learn More
Medchemexpress LLC Z-N-Me-Ala-OH | 21691-41-8 | MFCD00153386 | 99.4% | 237.25 g/mol | C12H15NO4 | 10 G

Supplier: Medchemexpress LLC HYW02244410G
Z-N-Me-Ala-OH is a Z-protected N-methyl alanine derivative supplied as a solid for research use. It has CAS 21691-41-8, molecular formula C12H15NO4, and a molecular weight of 237.25 g/mol. The product is offered at high reported purity for laboratory applications involving amino acid derivatives and peptide synthesis research.
- Z-protected N-methyl alanine derivative.
- High reported purity (~99.4%).
- Solid physical form for convenient storage and handling.
- Molecular weight 237.25 g/mol; formula C12H15NO4.
- Intended for research use only.
- Available in multiple pack sizes for lab-scale work.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.