Learn More
Medchemexpress LLC (Z)-Non-6-en-1-ol | 35854-86-5 | MFCD00015388 | 142.24 g·mol⁻¹ | C9H18O | 1 G

Supplier: Medchemexpress LLC HYW0175581G
(Z)-Non-6-en-1-ol (cis-6-nonen-1-ol) is an unsaturated primary alcohol supplied as a biochemical research reagent. It is intended for laboratory research use and has not been validated for medical applications.
- Chemical identity: (Z)-Non-6-en-1-ol; CAS 35854-86-5.
- Molecular formula C9H18O; molecular weight 142.24 g·mol⁻¹.
- Intended use: for research use only; not validated for medical applications.
- Storage recommendations: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
- Application category: biochemical assay reagent.
- Packaging: supplied in research-scale quantities suitable for laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.