Learn More
Medchemexpress LLC (Z)-ono 1301 | 153814-74-5 | 98.1% | 428.48 g/mol | C26H24N2O4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY106961A1MG
Encompass_Preferred
(Z)-ONO 1301 is the Z-isomer of a prostacyclin (PGI2) mimetic and long-acting prostacyclin agonist with reported thromboxane-synthase inhibitory activity. It is provided as a solid reference compound for use as an experimental control in pharmacology and signaling studies involving prostacyclin receptors, angiogenesis, and related assays.
  • Isomeric reference for prostacyclin agonist studies.
  • Exhibits prostacyclin agonist activity and thromboxane-synthase inhibition.
  • Supplied as an off-white to light-yellow solid suitable for analytical use.
  • High purity consistent with analytical grade reference material.
  • Molecular formula C26H24N2O4; molecular weight 428.48 g/mol.
Catalog No. 50-002-39890