Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Z)-Ono 1301 | 153814-74-5 | 98.1% | 428.48 g/mol | C26H24N2O4 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY106961A5MG

Encompass_Preferred

(Z)-ONO 1301 is the Z geometric isomer of the prostacyclin agonist ONO-1301, provided as a research reagent for use as an experimental control in studies of prostacyclin signaling and thromboxane synthase inhibition. The compound is supplied as a solid and characterized by COA/SDS data including molecular formula, molecular weight, and HPLC purity.

  • Used as an experimental control isomer in prostacyclin-related studies.
  • High purity, 98.1% by HPLC.
  • Molecular weight 428.48 g/mol.
  • Chemical formula C26H24N2O4.
  • Off-white to light yellow solid appearance.
  • Recommended storage: powder at -20°C; in solvent at -80°C.

Catalog No. 50-002-40588


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.